missing translation for 'onlineSavingsMsg'
Learn More

Influenza B Hemagglutinin Antibody (10A6E9C7), Biotin, Novus Biologicals™

Artikelnummer. 18612998 Alle ansehen Bio Techne Produkte
Change view
Click to view available options
Menge:
0.1 mL
Unit Size:
0.10 Milliliter
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Menge unitSize
18612998 0.1 mL 0.10 Milliliter
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18612998 Supplier Novus Biologicals Supplier No. NBP306570B

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Influenza B Hemagglutinin Monoclonal antibody specifically detects Influenza B Hemagglutinin in Virus samples. It is validated for ELISA, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen Influenza B Hemagglutinin
Anwendungen ELISA
Klassifikation Monoclonal
Klon 10A6E9C7
Konjugat Biotin
Verdünnung ELISA, Sandwich ELISA
Zusammensetzung PBS
Gen-Alias B1AG46
Wirtsspezies Mouse
Immunogen This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinantInfluenza B Hemagglutinin (ACA33493.1; Met 1-Ala 555).
Reinigungsverfahren Protein A purified
Menge 0.1 mL
Regulatorischer Status RUO
Primär oder sekundär Primary
Zielspezies Virus
Inhalt und Lagerung Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.