missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ QPCT Polyclonal Antibody
Sheep Polyclonal Antibody
Marke: Invitrogen™ PA5149191
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Reconstitute at 0.2 mg/mL in sterile PBS.
GPIP137 regulates the transport and translation of mRNAs of proteins involved in synaptic plasticity in neurons and cell proliferation/migration in multiple cell types. It binds directly and selectively to MYC and CCND2 RNAs. In neuronal cells, GPIP137 binds to several mRNAs associated with RNA granules including BDNF, CAMK2A, CREB1, MAP2, NTRK2, mRNAs, GRIN1, and KPNB1 mRNAs.
Spezifikation
| QPCT | |
| Polyclonal | |
| Unconjugated | |
| Qpct | |
| 5730422A13Rik; EC; GCT; Glutaminyl cyclase; glutaminyl-peptide cyclotransferase; glutaminyl-peptide cyclotransferase (glutaminyl cyclase); glutaminyl-tRNA cyclotransferase; Glutamyl cyclase; QC; Qpct; RGD1562284; similar to Glutaminyl-peptide cyclotransferase precursor (QC); sQC | |
| Sheep | |
| Antigen affinity chromatography | |
| RUO | |
| 25797 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
| Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| PBS with trehalose and no preservative | |
| Q16769 | |
| Qpct | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Glutaminyl-peptide Cyclotransferase/QPCT, Ala33-Leu361. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur