Learn More
Unity Lab Services™ Qualified Installation - USP/EP - Evolution Pro
Alle ansehen Thermo Scientific ProdukteDescription
- On-site installation of instrument
- Installation Qualification and Operational Qualification
- User orientation and instrument familiarization
- All engineer labor and travel charges for installation
Specifications
Specifications
| Zur Verwendung mit (Geräte) | Evolution Pro |
| Dauer | 1 Day |
| Produkttyp | Installation Service |
| Produktlinie | Thermo Scientific |
| Service Offering | Installation Qualification |