Learn More
Contec Quicktask Non-Magnetic Mop Frame, Stainless Steel
Alle ansehen Contec Produkte
Click to view available options
Autoklavierbar:
Packungsgröße:
1 Stück
Beschreibung
- Available in stainless steel or blue polypropylene
- Autoclavable
- Compatible with most common disinfectant solutions and solvents
- The QuickTask™ system consists of a variety of knitted polyester, polyester/cellulose, microfiber/foam, and microfiber flat mop heads that are easily installed onto a hinged mop head frame and handle assembly
- The QuickTask™ system is ideally suited for cleaning floors, including perforated cleanroom floors and grates
Spezifikation
Spezifikation
| Produkttyp | 1 Piece Lay Flat Frame |
| Sterilität | Non-sterile |
| Autoklavierbar | Autoclavable |
| Einwegprodukt | Reusable |
| Color-coded | No |
| Material | Stainless Steel |