Learn More
R&D Systems™ Rat V1b Vasopressin R/AVPR1B (NP_058901) VersaClone cDNA
Artikelnummer. 18481860
Ansicht ändern
Click to view available options
Menge:
10 μg
Packungsgröße:
10 Mikrogramm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge Packungsgröße
18481860 10 μg 10 Mikrogramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Artikelnummer. 18481860 Lieferant R&D Systems™ Lieferanten-Nr. RDC0788
  • ONLINE EXCLUSIVE PRICE
    341.00 €
    322.35 € / 10 Mikrogramm
    Sparen 18.65 €
    5% Rabatt
 Eingeschränkter Artikel
Verfügbar ab: 02-12-2026
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
missing translation for 'validMainframeMsg'
Arginine Vasopressin Receptor 1B
The Rat V1b Vasopressin R/AVPR1B (NP_058901) VersaClone cDNA clone contains the complete ORF for V1b Vasopressin R/AVPR1B, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The V1b Vasopressin R/AVPR1B insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
Bestellinformation
Ships at ambient temperature.
TRUSTED_SUSTAINABILITY
Spezifikation
Gen rAVPR1B
Zugriffsnummer NP_058901
Konzentration 10 μg at 0.2 μg/μL
Zusammensetzung cDNA is provided in 10mM Tris-Cl, pH 8.5
Einsatztyp 1277bp
Inhalt und Lagerung Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Spezies Ratte
Produktlinie R&D Systems
Menge 10 μg
For Reasearch Use Only; Not for Use in Humans