missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(R)-GNE-140, MedChemExpress
Description
(R)-GNE-140 is a potent lactate dehydrogenase A (LDHA) inhibitor, with IC50s of 3 nM and 5 nM for LDHA and LDHB, respectively
Specifications
Specifications
| Chemischer Name oder Material | (R)-GNE-140 |
| Physikalische Form | Solid |
| CAS | 2003234-63-5 |
| Farbe | Yellow |
| Menge | 10 mg |
| Hinweise zur Reinheitsqualität | Research |
| Summenformel | C25H23ClN2O3S2 |
| Zur Verwendung mit (Anwendung) | Cancer-programmed cell death |
| Löslichkeitsinformationen | DMSO : ≥ 50 mg/mL (100.19 mM) |
| SMILES | O=C1N[C@@](C2=CSC=C2)(C3=CC=C(N4CCOCC4)C=C3)CC(O)=C1SC5=CC=CC=C5Cl |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?