Learn More
(R,S)-2,2-Dimethyl-1,3-dioxolane-4-methanol, TRC
Artikelnummer. 30008707
Change view
Click to view available options
Menge:
50 g
100 g
250 g
Packungsgröße:
100 Gramm
250 Gramm
50 Gramm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
30008707 250 g 250 Gramm
30012307 50 g 50 Gramm
30008810 100 g 100 Gramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30008707 Lieferant Toronto Research Chemicals Lieferanten-Nr. TRCD472640250G
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemische Identifikatoren
CAS 100-79-8
Summenformel C6H12O3
Molekulargewicht (g/mol) 132.16
Synonym (RS)-Solketal, (+/-)-1,2-O-Isopropylideneglycerol, (+/-)-Glycerol 1,2-Acetonide, 1,2-Isopropylideneglycerin, 1,2-Isopropylideneglycerol, 2,3-(Isopropylidenedioxy)propanol, 2,3-Isopropylideneglycerol, Acetone Glyceryl Acetal, Dioxolan, α,β-Isopropylidene Ether, Glycerolacetone, NSC 59720;
SMILES CC1(C)OCC(CO)O1
Spezifikation
Chemischer Name oder Material (R,S)-2,2-Dimethyl-1,3-dioxolane-4-methanol
Menge 250 g
InChI-Formel InChI=1S/C6H12O3/c1-6(2)8-4-5(3-7)9-6/h5,7H,3-4H2,1-2H3
Formelmasse 132.0786