missing translation for 'onlineSavingsMsg'
Learn More

(R,S)-Anabasin-2,4,5,6-d4, TRC

Product Code. 30346450
Change view
Click to view available options
Menge:
5 mg
Unit Size:
5 Milligramm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Menge Unit Size
30346450 5 mg 5 Milligramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30346450 Supplier Toronto Research Chemicals Supplier No. TRCA6371805MG
 Eingeschränkter Artikel
Verfügbar ab: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Hochreine organische Moleküle und analytische Standards, strategisch weltweit geliefert, um Innovation und wirtschaftlichen Erfolg zu ermöglichen.

TRUSTED_SUSTAINABILITY

Specifications

Chemischer Name oder Material (R,S)-Anabasine-2,4,5,6-d4
CAS 1020719-08-7
Hinweise zur Reinheitsqualität HPLC
Summenformel C10 2H4 H10 N2
Menge 5 mg
Synonym rac-Anabasine-D4 (pyridine-D4), (R,S)-Anabasine-2,4,5,6-d4
InChI-Formel InChI=1S/C10H14N2/c1-2-7-12-10(5-1)9-4-3-6-11-8-9/h3-4,6,8,10,12H,1-2,5,7H2/i3D,4D,6D,8D
SMILES [2H]c1nc([2H])c(C2CCCCN2)c([2H])c1[2H]
IUPAC-Name 2,3,4,6-Tetradeuterio-5-piperidin-2-ylpyridin
Molekulargewicht (g/mol) 166.26
Formelmasse 166.1408
Reinheit (%) >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.