Beschreibung
Rab7a Monoclonal antibody specifically detects Rab7a in Human samples. It is validated for Western Blot, ELISA
Spezifikation
Spezifikation
| Antigen | Rab7a |
| Anwendungen | Western Blot, ELISA |
| Klassifikation | Monoclonal |
| Klon | 100000 |
| Konjugat | HRP |
| Zusammensetzung | PBS |
| Gen-Alias | CMT2B, FLJ20819, member RAS oncogene family, PSN, RAB7A, member RAS oncogene family, Ras-associated protein RAB7, ras-related protein Rab-7a |
| Wirtsspezies | Mouse |
| Immunogen | Recombinant human RAB7A (1-207aa) purified from E. coli |
| Reinigungsverfahren | Protein A purified |
| Mehr anzeigen |