Learn More
ADRA1D Rabbit anti-Bovine, Dog, Horse, Human, Monkey, Pig, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of ADRA1D.
Marke: Abnova PAB25531.50ug
Beschreibung
Alpha-1-adrenergic receptors (alpha-1-ARs) are members of the G protein-coupled receptor superfamily. They activate mitogenic responses and regulate growth and proliferation of many cells. There are 3 alpha-1-AR subtypes: alpha-1A, -1B and -1D, all of which signal through the Gq/11 family of G-proteins and different subtypes show different patterns of activation. This gene encodes alpha-1D-adrenergic receptor. Similar to alpha-1B-adrenergic receptor gene, this gene comprises 2 exons and a single intron that interrupts the coding region. [provided by RefSeq
Spezifikation
| ADRA1D | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of ADRA1D. | |
| In PBS (0.09% sodium azide) | |
| ADRA1/ADRA1A/ADRA1R/ALPHA1/DAR/dJ779E11.2 | |
| Rabbit | |
| Immunoaffinity chromatography | |
| RUO | |
| 146 | |
| Bovine, Canine, Equine, Human, Monkey, Pig, Rabbit | |
| Liquid |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1-3 ug/mL) The optimal working dilution should be determined by the end user. | |
| ADRA1D | |
| ADRA1D | |
| A synthetic peptide corresponding to 20 amino acids at 3rd cytoplasmic domain of human ADRA1D. | |
| 50 μg | |
| Primary | |
| BLAST analysis of the peptide immunogen showed no homology with other human proteins. | |
| Store at 4°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.
For Research Use Only