Learn More
APH1A Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of APH1A.
Marke: Abnova PAB4887.400uL
Beschreibung
APH1 is a multipass transmembrane protein that interacts with presenilin (see PSEN1; MIM 104311) and nicastrin (APH2; MIM 605254) as a functional component of the gamma-secretase complex. The gamma-secretase complex is required for the intramembrane proteolysis of a number of membrane proteins, including the amyloid-beta precursor protein (APP; MIM 104760) and Notch (MIM 190198).[supplied by OMIM
Spezifikation
| APH1A | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of APH1A. | |
| In PBS (0.09% sodium azide) | |
| NP_001071096;NP_057106;Q96BI3 | |
| APH1A | |
| A synthetic peptide (conjugated with KLH) corresponding to N-terminus of human APH1A. | |
| 400 μL | |
| Primary | |
| Human | |
| Liquid |
| ELISA, Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| ELISA (1:1000) Western Blot (1:100-500) Immunohistochemistry (1:50-100) The optimal working dilution should be determined by the end user. | |
| APH1A | |
| 6530402N02Rik/APH-1A/CGI-78 | |
| Rabbit | |
| Protein G purification | |
| RUO | |
| 51107 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only