Learn More
Invitrogen™ Rabbit anti-Bovine IgG (H+L) Secondary Antibody
Rabbit Polyclonal Secondary Antibody
Brand:  Invitrogen™ SA510310
Product Code. 17139685
  • 63.25 € / 1 Milligramm
om dit product te kopen Registreer vandaag om een webaccount aan te maken
This item is not returnable. View return policy
Description
Description
This antibody will react with bovine IgG and with light chains common with other bovine immunoglobulins as determined by ELISA and IEP techniques. This antibody may cross-react with IgG from other species.
Anti-Bovine secondary antibodies are affinity-purified antibodies with well-characterized specificity for bovine (cow, ox) immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Bovine IgG (H+L)
Polyclonal
Unconjugated
Rabbit
Antigen affinity chromatography
RUO
Bovine
Secondary Antibody
IgG
ELISA, Immunoelectrophoresis, Immunohistochemistry, Western Blot
1 mg/mL
PBS with 0.09% sodium azide; pH 7.2
Bovine IgG h+l.
1 mg
Secondary
4°C
Liquid
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications