Learn More
CAMK2A/CAMK2B/CAMK2D (phospho T305), Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic phosphopeptide of human CAMK2A/CAMK2B/CAMK2D.
Marke: Abnova PAB29254.100uL
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Spezifikation
| CAMK2A/CAMK2B/CAMK2D | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic phosphopeptide of human CAMK2A/CAMK2B/CAMK2D. | |
| In PBS (without mg2+ and Ca2+), 150mM NaCl, pH 7.4 (50% glycerol, 0.02% sodium azide) | |
| CAMKA/KIAA0968 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 815/816/817 | |
| Human, Mouse, Rat | |
| Liquid |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1:50∼1:100) Western Blot (1:500∼1:1000) The optimal working dilution should be determined by the end user. | |
| Q9UQM7 | |
| CAMK2A | |
| Synthetic phosphopeptide (conjugated with KLH) corresponding to residues surrounding T305 of human CAMK2A/CAMK2B/CAMK2D. | |
| 100 μL | |
| Primary | |
| CAMK2A/CAMK2B/CAMK2D (phospho T305) polyclonal antibody detects endogenous levels of human CAMK2A/CAMK2B/CAMK2D only when phosphorylated at threonine 305. | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |