Learn More
dcr-1, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of dcr-1.
Marke:  Abnova PAB3792.400uL
Artikelnummer. 16088797
  • 384.00 € / 400 Mikroliter
om dit product te kopen Registreer vandaag om een webaccount aan te maken
Spezifikation
dcr-1
Polyclonal
Rabbit polyclonal antibody raised against synthetic peptide of dcr-1.
In PBS (0.09% sodium azide)
NP_498761;P34529
Rabbit
Protein G purification
RUO
176138
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
ELISA, Western Blot
Unconjugated
Western Blot (1:1000) The optimal working dilution should be determined by the end user.
dcr-1
dcr-1
A synthetic peptide (conjugated with KLH) corresponding to internal region of C.elegans dcr-1.
400 μL
Primary
Nematode
Liquid
Produktvorschläge
Produktvorschläge
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Dokumentation
Zertifikate
Aktionen
Aktionen