Learn More
GAB1 (phospho Y659), Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic phosphopeptide of GAB1.
Marke: Abnova PAB16972.100ug
Beschreibung
The protein encoded by this gene is a member of the IRS1-like multisubstrate docking protein family. It is an important mediator of branching tubulogenesis and plays a central role in cellular growth response, transformation and apoptosis. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq
Spezifikation
| GAB1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic phosphopeptide of GAB1. | |
| In 20mM PBS, 0.15M NaCl, pH 7.2 (0.01% sodium azide) | |
| Q13480 | |
| Rabbit | |
| 100 μg | |
| Primary | |
| This antibody detects endogenous levels of GAB1 only when phosphorylated at tyrosine 659. | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Western Blot (1:500-1:1000) Immunohistochemistry (1:50-1:100) ELISA (1:4000) The optimal working dilution should be determined by the end user. | |
| GAB1 | |
| GAB1 | |
| Synthetic phosphopeptide corresponding to residues surrounding Y659 of human GAB1. | |
| RUO | |
| 2549 | |
| Human, Mouse, Rat | |
| Liquid |