Learn More
Invitrogen™ Rabbit anti-Human IgA (Heavy chain) Secondary Antibody
Rabbit Polyclonal Secondary Antibody
Marke: Invitrogen™ PA516388
Beschreibung
PA5-16388 targets Human IgA (alpha-Heavy Chain) in IHC (P), WB and FACS applications and shows reactivity with Human samples. This antibody is not recommended for Daudi cells in FACS applications. This antibody detects human and mouse IgA in Western blot. The PA5-16388 immunogen is igA isolated from a pool of normal human sera.
Anti-Human secondary antibodies are affinity-purified antibodies with well-characterized specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Spezifikation
| Human IgA (Heavy chain) | |
| Polyclonal | |
| PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4 | |
| IgA isolated from a pool of normal human sera. | |
| 500 μL | |
| Secondary | |
| 4°C | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Rabbit | |
| Ig fraction | |
| RUO | |
| Human | |
| Secondary Antibody | |
| IgG |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.