missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rabbit anti-Human IgG (H+L) Secondary Antibody, HRP (Pre-adsorbed), Novus Biologicals™
Rabbit Polyclonal Antibody
Marke: Novus Biologicals NB120-6759
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
IgG (H+L) Polyclonal specifically detects IgG (H+L) in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry.
Spezifikation
| IgG (H+L) | |
| Polyclonal | |
| Western Blot 1:1000 - 1:5000, ELISA 1:50000 - 1:100000, Immunohistochemistry 1:500 - 1:2500 | |
| Rabbit anti-Human IgG (H+L) Secondary Antibody [HRP] (Pre-adsorbed) was produced by repeated immunization with human IgG whole molecule in rabbit. | |
| 1 mg | |
| Adaptive Immunity, Immunology | |
| This antibody was pre-adsorbed against Mouse Serum Proteins. No reaction was observed against Mouse Serum Proteins. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| HRP | |
| Rabbit | |
| Multi-step | |
| RUO | |
| Secondary | |
| Human | |
| Purified |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur