missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MER2 (phospho S30), Rabbit anti-Yeast, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic phosphopeptide of MER2.
Marke: Abnova PAB11308.100ug
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Spezifikation
| MER2 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic phosphopeptide of MER2. | |
| In 20mM KH2PO4, 150mM NaCl, pH 7.2 (0.01% sodium azide) | |
| MER2 | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Reactivity occurs against Saccharomyces cerevisiae Mer2 pS30 protein and This antibody is specific to the phosphorylated form of the protein. Reactivity with non-phosphorylated Saccharomyces cerevisiae Mer2 is minimal by ELISA and western blot. | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Immunoprecipitation, Western Blot | |
| Unconjugated | |
| ELISA (1:5000-1:25000) Western Blot (1:1000-1:10000) The optimal working dilution should be determined by the end user. | |
| REC107 | |
| REC107 | |
| Synthetic phosphopeptide corresponding to residues surrounding S30 of Saccharomyces cerevisiae MER2. | |
| RUO | |
| 853478 | |
| Yeast | |
| Liquid |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur