Learn More
Rabbit anti-Mouse Kappa Light Chain Secondary Antibody, PE, Novus Biologicals™
Rabbit Polyclonal Antibody
Marke: Novus Biologicals NBP1-72791PE
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Kappa Light Chain Polyclonal antibody specifically detects Kappa Light Chain in Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry
Spezifikation
| Kappa Light Chain | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Multi-step | |
| RUO | |
| Secondary | |
| Mouse | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry | |
| PE | |
| HCAK1, IGKCD, immunoglobulin kappa constant, Km, MGC111575, MGC62011, MGC72072, MGC88770, MGC88771, MGC88809 | |
| Mouse kappa light chain | |
| 0.5 mL | |
| Adaptive Immunity, Immunology | |
| 3514 | |
| Store at 4°C in the dark. |