missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAD9 Rabbit anti-Yeast, Polyclonal Antibody, Abnova™
Spezifikation
Spezifikation
| Antigen | RAD9 |
| Anwendungen | ELISA, Western Blot |
| Klassifikation | Polyclonal |
| Konjugat | Unconjugated |
| Beschreibung | Rabbit polyclonal antibody raised against synthetic peptide of RAD9. |
| Verdünnung | Sandwich ELISA (1:5000) Western Blot (1:100-1:500) The optimal working dilution should be determined by the end user. |
| Zusammensetzung | In 20mM KH2PO4, 150mM NaCl, pH 7.2 (0.01% sodium azide) |
| Gen | RAD9 |
| Gensymbole | RAD9 |
| Wirtsspezies | Rabbit |
| Mehr anzeigen |
For Research Use Only
Name des Produkts
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?