missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RAC1 Recombinant Rabbit Monoclonal Antibody (PSH05-09)

Rabbit Recombinant Monoclonal Antibody
Marke: Invitrogen™ MA556402
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
RAC1 Recombinant Monoclonal Antibody for Western Blot, ICC/IF
The protein encoded by this gene is a GTPase which belongs to the RAS superfamily of small GTP-binding proteins. Members of this superfamily appear to regulate a diverse array of cellular events, including the control of cell growth, cytoskeletal reorganization, and the activation of protein kinases. Two transcript variants encoding different isoforms have been found for this gene.
Spezifikation
| RAC1 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with 0.1% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| P63000 | |
| Rac1 | |
| Recombinant protein within human RAC1 aa 1-192/192. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Western Blot, Immunocytochemistry | |
| PSH05-09 | |
| Unconjugated | |
| Rac1 | |
| AL023026; cell migration-inducing gene 5 protein; D5Ertd559e; GTPase; MGC111543; MIG5; p21-Rac1; Rac 1b; Rac family small GTPase 1; RAC1; Rac-1; Rac2; rac2 GTP-binding protein; ras-like protein TC25; RAS-related C3 botulinum substrate 1; ras-related C3 botulinum toxin substrate 1; ras-related C3 botulinum toxin substrate 1 (rho family small GTP binding protein Rac1); ras-related C3 botulinum toxin substrate 1 (rho family, small GTP binding protein Rac1); rho family, small GTP binding protein Rac1; TC25; TC-25 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 5879 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur