Learn More
RAD54L Antibody (Rad54 4E3/1), Alexa Fluor™ 700, Novus Biologicals™
Alle ansehen Bio Techne ProdukteBeschreibung
RAD54L Monoclonal specifically detects RAD54L in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Spezifikation
Spezifikation
| Antigen | RAD54L |
| Anwendungen | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence |
| Klassifikation | Monoclonal |
| Klon | Rad54 4E3/1 |
| Konjugat | Alexa Fluor 700 |
| Verdünnung | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence |
| Zusammensetzung | 50mM Sodium Borate with 0.05% Sodium Azide |
| Gen-Alias | EC 3.6.1, hHR54EC 3.6.4.-, HR54, hRAD54HRAD54, RAD54 (S.cerevisiae)-like, RAD54 homolog, RAD54ADNA repair and recombination protein RAD54-like, RAD54-like (S. cerevisiae) |
| Gensymbole | RAD54L |
| Wirtsspezies | Mouse |
| Mehr anzeigen |