Learn More
Rad9 Antibody (OTI7B11), DyLight 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-73783UV
Product Code. 18150857
  • 751.00 € / 0.10 Milliliter
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
This item is not returnable. View return policy
Description
Description
Rad9 Monoclonal specifically detects Rad9 in Human samples. It is validated for Western Blot, Immunohistochemistry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Rad9
Monoclonal
DyLight 350
50mM Sodium Borate with 0.05% Sodium Azide
RAD9A
Full-length protein expressed in 293T cell transfected with human Rad9 expression vector.
0.1 mL
Primary
Human
IgG2a
Western Blot, Immunohistochemistry
OTI7B11
Western Blot, Immunohistochemistry
cell cycle checkpoint control protein RAD9A, DNA repair exonuclease rad9 homolog A, EC 3.1.11.2, hRAD9, RAD9, RAD9 (S. pombe) homolog, RAD9 homolog A (S. pombe)
Mouse
Affinity Purified
Apoptosis, Cancer, Checkpoint signaling, Chromatin Research, DNA Repair, Phospho Specific, Tumor Suppressors
5883.0
Store at 4°C in the dark.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications