missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Radixin Recombinant Rabbit Monoclonal Antibody (24GB1420)
GREENER_CHOICE

Artikelnummer. p-200110644
Change view
Click to view available options
Menge:
20 μL
100 μL
Packungsgröße:
100 Mikroliter
20 Mikroliter
Artikelnummer. Menge unitSize
30275563 100 μL 100 Mikroliter
30276278 20 μL 20 Mikroliter
2 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Artikelnummer. 30275563

Marke: Invitrogen™ MA557674

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Rabbit Recombinant Monoclonal Antibody

Radixin Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow

Radixin is a cytoskeletal protein that may be important in linking actin to the plasma membrane. It is highly similar in sequence to both ezrin and moesin. The radixin gene has been localized by fluorescence in situ hybridization to 11q23. A truncated version representing a pseudogene (RDXP2) was assigned to Xp21.3. Another pseudogene that seemed to lack introns (RDXP1) was mapped to 11p by Southern and PCR analyses. Multiple alternatively spliced transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq, May 2012].
TRUSTED_SUSTAINABILITY

Spezifikation

Antigen Radixin
Anwendungen Flow Cytometry, Western Blot, Immunocytochemistry
Klassifikation Recombinant Monoclonal
Klon 24GB1420
Konjugat Unconjugated
Zusammensetzung PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gen Rdx
Gen-Zugriffsnummer P26043, P35241
Gen-Alias AA516625; DFNB24; ESP10; radixin; radixin isoform; Rdx
Gensymbole Rdx
Wirtsspezies Rabbit
Immunogen A synthesized peptide derived from human Radixin (aa 350-430).
Reinigungsverfahren Affinity Chromatography
Menge 100 μL
Regulatorischer Status RUO
Primär oder sekundär Primary
Gen-ID (Entrez) 19684, 315655, 5962
Zielspezies Human, Mouse, Rat
Inhalt und Lagerung -20°C
Produkttyp Antibody
Form Liquid
Isotype IgG
Mehr anzeigen Weniger anzeigen
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.