missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RALA Mouse anti-Human, Monkey, Mouse, Clone:4G8C7, Abnova™
Mouse monoclonal antibody raised against recombinant human RALA.
Marke: Abnova MAB16743.100ug
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
The product of this gene belongs to the small GTPase superfamily, Ras family of proteins. GTP-binding proteins mediate the transmembrane signaling initiated by the occupancy of certain cell surface receptors. This gene encodes a low molecular mass ras-like GTP-binding protein that shares about 50% similarity with other ras proteins. [provided by RefSeq]Spezifikation
| RALA | |
| Monoclonal | |
| Unconjugated | |
| ELISA (1:10000) Western Blot (1:500-1:2000) Immunohistochemistry (1:200-1:1000) Immunocytochemistry (1:200-1:1000) Flow Cytometry (1:200-1:400) The optimal working dilution should be determined by the end user. |
|
| RALA | |
| Mouse | |
| 100 μg | |
| Human, Monkey, Mouse | |
| Antibody | |
| IgG1 |
| ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
| 4G8C7 | |
| v-ral simian leukemia viral oncogene homolog A (ras related) | |
| In PBS (0.05% sodium azide) | |
| MGC48949, RAL | |
| Recombinant protein corresponding to amino acid 71-203 of human RALA from E. coli. | |
| 5898 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| Liquid |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur