Learn More
Invitrogen™ RAP1A/RAP1B Recombinant Rabbit Monoclonal Antibody (24GB1030)
Beschreibung
RAP1A/RAP1B Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
Spezifikation
Spezifikation
| Antigen | RAP1A/RAP1B |
| Anwendungen | Flow Cytometry, Western Blot, Immunocytochemistry |
| Klassifikation | Recombinant Monoclonal |
| Klon | 24GB1030 |
| Konjugat | Unconjugated |
| Zusammensetzung | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gen | RAP1A |
| Gen-Zugriffsnummer | P61224, P62834, P62835, P62836, Q62636, Q99JI6 |
| Gen-Alias | 2810443E11Rik; AI848598; C21KG; G-22K; GTP-binding protein smg p21A; GTP-binding protein smg p21B; GTP-binding protein SMG-P21A; K-REV; KREV1; Krev-1; OK/SW-cl.11; RAL1B; rap 1A; RAP1; RAP1A; RAP-1A; RAP1A, member of RAS oncogene family; Rap1b; RAP1B, member of RAS oncogene family; Ras family small GTP binding protein RAP1B; RAS related protein 1b; RAS-related protein 1a; Ras-related protein Krev-1; Ras-related protein Rap-1A; RAS-related protein RAP1B; ras-related protein Rap-1b; RAS-related protein-1a; small GTP binding protein; SMGP21 |
| Gensymbole | RAP1A, RAP1B |
| Mehr anzeigen |