Learn More
Invitrogen™ RAP1A Recombinant Rabbit Monoclonal Antibody (HL2415)
GREENER_CHOICE
Rabbit Recombinant Monoclonal Antibody
Marke:  Invitrogen™ MA556854
Artikelnummer. 30275823
  • 514.00 € / 100 Mikroliter
om dit product te kopen Registreer vandaag om een webaccount aan te maken
Beschreibung
Beschreibung
RAP1A Recombinant Monoclonal Antibody for Western Blot
The product of this gene belongs to the family of RAS-related proteins. These proteins share approximately 50% amino acid identity with the classical RAS proteins and have numerous structural features in common. The most striking difference between RAP proteins and RAS proteins resides in their 61st amino acid: glutamine in RAS is replaced by threonine in RAP proteins. The product of this gene counteracts the mitogenic function of RAS because it can interact with RAS GAPs and RAF in a competitive manner. Two transcript variants encoding the same protein have been identified for this gene.
TRUSTED_SUSTAINABILITY
Spezifikation
Spezifikation
RAP1A
Recombinant Monoclonal
1 mg/mL
PBS with no preservative
P62834
RAP1A
Recombinant fragment of human RAP1A.
100 μL
Primary
Human
Antibody
IgG
Western Blot
HL2415
Unconjugated
RAP1A
AI848598; C21KG; G-22K; GTP-binding protein; GTP-binding protein smg p21A; GTP-binding protein SMG-P21A; KREV1; Krev-1; rap 1A; RAP1; RAP1A; RAP-1A; RAP1A, member of RAS oncogene family; RAS-related protein 1a; ras-related protein Krev-1; Ras-related protein Rap-1A; RAS-related protein-1a; SMGP21
Rabbit
Protein A
RUO
5906
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Produktvorschläge
Produktvorschläge
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Dokumentation
Zertifikate
Aktionen
Aktionen