Learn More
Thermo Scientific™ Rapid DNA Ligation Kit Promotion
Alle ansehen Thermo Scientific ProdukteBeschreibung
Rapid DNA Ligation Kit enables fast sticky-end or blunt-end DNA ligation in only 5 minutes at room temperature.
The kit contains T4 DNA ligase and a specially-formulated 5X rapid ligation buffer optimized for fast and efficient DNA ligation. Fast ligation efficiency is equal to that obtained with T4 DNA ligase in a standard 1 hour ligation. The ligation reaction mixture can be used directly for bacterial transformation with the TransformAid Bacterial Transformation Kit or with other conventional transformation procedures.
Highlights
- Fast -ligation of either sticky- or blunt-end DNA in only 5 minutes
- Convenient -reaction mixture can be used directly for bacterial transformation
Spezifikation
Spezifikation
| Zur Verwendung mit (Anwendung) | Routine cloning experiments; Blunt-end cloning; Library construction; TA cloning. |
| Anzahl Reaktionen | 150 |
| Produkttyp | DNA-Schnell-Ligationskit |
| Menge | 150 Reaktionen |