Learn More
Raptor Rabbit anti-Human, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 20984-1-AP-150UL
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
The antibody is specific to RPTOR.
Regulatory associated protein of mTOR (RAPTOR) is a binding partner of target of rapamycin (TOR). It binds to 4EBP1 and p70alpha. The binding of raptor to mTOR is essential for the mTOR-catalyzed phosphorylation of 4EBP1 in vitro. It enhances the mTOR kinase activity toward p70alpha. It is also an essential scaffold for the mTOR-catalyzed phosphorylation of 4EBP1 and mediates TOR action in vivo.Spezifikation
| Raptor | |
| Polyclonal | |
| Unconjugated | |
| RPTOR | |
| KIAA1303, raptor, RPTOR | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 287871, 57521 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.2 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8N122 | |
| Rptor | |
| Peptide | |
| 150 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |