Learn More
Ahsa1, Rat, Clone: 25F2.D10, Abnova™
Rat monoclonal antibody raised against Ahsa1.
Marke: Abnova MAB6669.100ug
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Spezifikation
| Ahsa1 | |
| Monoclonal | |
| Unconjugated | |
| Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
| Ahsa1 | |
| Ahsa1 | |
| Mouse Aha1. | |
| RUO | |
| 217737 | |
| Human, Mouse, Rat | |
| Liquid |
| ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blot | |
| 25F2.D10 | |
| Rat monoclonal antibody raised against Ahsa1. | |
| In 20mM potassium phosphate buffer, 150mM NaCl, pH 7.2 | |
| BC023857/MGC36589/MGC36618/p38 | |
| Rat | |
| 100 μg | |
| Primary | |
| Approximately 38 KDa, can run at up to 45 KDa on SDS-PAGE. | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG2a κ |