missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rat anti-Mouse IgG (H+L) Secondary Antibody, Invitrogen™
Rat Monoclonal Secondary Antibody
Marke: Invitrogen SA135633
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Thermo Scientific Anti-Mouse secondary antibodies are affinity-purified antibodies with well-characterized specificity for mouse immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Spezifikation
| Mouse IgG (H+L) | |
| Monoclonal | |
| 1 mg/mL | |
| Rat | |
| Purified | |
| RUO | |
| Mouse | |
| Liquid |
| ELISA, Immunohistochemistry (Frozen) | |
| LO-MG-7 | |
| Unconjugated | |
| Immunoglobulins. | |
| 1 mg | |
| Secondary | |
| 4° C | |
| IgM κ |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur