Learn More
Rat anti-Mouse IgG2b, Clone: R12-3, FITC, Secondary Antibody, BD
Marke: BD Biosciences 553395
Beschreibung
The R12-3 antibody recognizes an epitope in the CH3 domain of mouse IgG2b of Igh-C[a] and Igh-C[b] haplotypes. It does not react with other Ig isotypes. Detection of surface immunoglobulin on B lymphoma cells has been demonstrated with R12-3 mAb. This ant
Host Species: Rat
Clone: R12-3
Isotype: IgG2a, κ
Species Reactivity: Mouse
Immunogen: Pooled Mouse Ig
ELISA, Flow Cytometry
Spezifikation
| Rat anti-Mouse IgG2b | |
| R12-3 | |
| FITC | |
| 0.5 mg | |
| Immunology | |
| Mouse |
| Monoclonal | |
| 0.5mg/mL | |
| Rat | |
| RUO | |
| Secondary | |
| IgG2a κ |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.