missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Rat IgM Isotype Control (eBRM), Biotin, eBioscience™

Artikelnummer. 15209809
Ansicht ändern
Click to view available options
Menge:
50 μg
Packungsgröße:
50 Mikrogramm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
15209809 50 μg 50 Mikrogramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 15209809 Lieferant Invitrogen™ Lieferanten-Nr. 13434181

Please to purchase this item. Need a web account? Register with us today!

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Rat Monoclonal Isotype Control

The monoclonal rat IgM, kappa is useful as an isotype control immunoglobulin. Applications Reported: Rat IgM Isotype Control Biotin has been reported for use in flow cytometric analysis and ELISA. Applications Tested: Rat IgM Isotype Control has been tested by flow cytometric analysis of normal human peripheral blood cells and mouse spleen cells. It should be used at the same concentration as the experimental antibody. Filtration: 0.2 μm post-manufacturing filtered.

Rat IgM is an immunoglobulin M isotype antibody produced by B cells in rats. It plays a crucial role in the immune system by recognizing and neutralizing antigens, such as bacteria and viruses. Rat IgM has been extensively studied and is widely used in research applications, including immunology, microbiology, and biotechnology. It is commonly used as a control in experiments, as well as a diagnostic tool for detecting infectious diseases and autoimmune disorders. Researchers have developed various techniques for producing and purifying Rat IgM, including hybridoma technology, protein A/G chromatography, and affinity chromatography. Rat IgM is a valuable tool in the field of immunology, and its use has contributed to a better understanding of the immune system and the development of new treatments for various diseases.
TRUSTED_SUSTAINABILITY

Spezifikation

Antigen Rat IgM
Anwendungen Control, Flow Cytometry
Klassifikation Regelung
Klon eBRM
Konzentration 0.5 mg/mL
Konjugat Biotin
Zusammensetzung PBS with 0.09% sodium azide; pH 7.2
Gen-Alias Igh-6; IgM; immunoglobulin heavy chain 6; RGD1359202
Wirtsspezies Rat
Reinigungsverfahren Affinity chromatography
Menge 50 μg
Regulatorischer Status RUO
Primär oder sekundär Isotype Control (Primary)
Zielspezies Not Applicable
Inhalt und Lagerung 4°C, store in dark, DO NOT FREEZE!
Produkttyp Isotype Control
Form Liquid
Isotype IgM κ
Mehr anzeigen Weniger anzeigen
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.