Learn More
EduLab™ Ray Box
Artikelnummer. 13152920
Artikelnummer. 13152920 Lieferant EduLab™ Lieferanten-Nr. EDU1207
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Alternative Produkte anzeigen
missing translation for 'validMainframeMsg'
Used for a wide range of optics experiments.
  • Designed with two slots to hold blanking plates, seven grooves for lenses
  • Supplied with 12V, 20 - 24W, bulb, holder, slit plates, blanking plates and instructions
Spezifikation
Enthält 12V, 20 - 24W, bulb, holder, slit plates, blanking plates and instructions
Produkttyp Ray Box