missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RBCK1 Antibody (CL4289) [HRP], Novus Biologicals™
Mouse Monoclonal Antibody
Marke: Novus Biologicals NBP2-59048H
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
RBCK1 Monoclonal antibody specifically detects RBCK1 in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin)
Spezifikation
| RBCK1 | |
| Monoclonal | |
| HRP | |
| C20orf18, EC 6.3.2.-, HBV associated factor 4, HBV-associated factor 4, Heme-oxidized IRP2 ubiquitin ligase 1, Hepatitis B virus X-associated protein 4, HOIL-1, RanBP-type and C3HC4-type zinc finger containing 1, ranBP-type and C3HC4-type zinc finger-containing protein 1, RBCC protein interacting with PKC1, RBCK2, RNF54RING finger protein 54, UBCE7IP3HOIL1, ubiquitin conjugating enzyme 7 interacting protein 3, Ubiquitin-conjugating enzyme 7-interacting protein 3, XAP3, XAP4chromosome 20 open reading frame 18, ZRANB4 | |
| This antibody was developed using a recombinant protein derived from Q9BYM8, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Zinc Finger | |
| 10616 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| CL4289 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction