Learn More
REC8 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 10793-1-AP-150UL
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
This antibody is a rabbit polyclonal antibody raised against residues near the C terminus of human REC8. This antibody can recognize the monomeric, non-phosphorylated form (63-70 kDa) of REC8 and other forms(80-100 kDa) which represent either post transla
This gene encodes a member of the kleisin family of SMC (structural maintece of chromosome) protein partners. The protein localizes to the axial elements of chromosomes during meiosis in both oocytes and spermatocytes. In the mouse, the homologous protein is a key component of the meiotic cohesion complex, which regulates sister chromatid cohesion and recombination between homologous chromosomes. Multiple alternatively spliced variants, encoding the same protein, have been found for this gene.Spezifikation
| REC8 | |
| Polyclonal | |
| Unconjugated | |
| REC8 | |
| Cohesin Rec8p, HR21spB, REC8, REC8 homolog (yeast), REC8L1, Rec8p | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 56739, 9985 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.21 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O95072, Q8C5S7 | |
| Rec8 | |
| REC8 Fusion Protein Ag1250 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |