missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Bovine Angiogenin-1 Protein
Click to view available options
Menge:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Packungsgröße:
1 Milligramm
10 Mikrogramm
100 Mikrogramm
200 Mikrogramm
50 Mikrogramm
500 Mikrogramm
Specifications
Specifications
| Name | Angiogenin-1 Protein |
| Menge | 10 μg |
| Kennzeichnung | Research Use Only |
| Endotoxin-Konzentration | Not determined. |
| Gensymbol | ANG1 |
| Proteinlänge | Full Length |
| Produkttyp | Recombinant Protein |
| Kreuzreaktivität | Bovine |
| Spezies | E. coli |
| Proteinmarkierung | His |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction