missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Cynomolgus B7-H6 Fc Chimera Protein

Artikelnummer. 18778947
Ansicht ändern
Click to view available options
Menge:
50 μg
Packungsgröße:
50 Mikrogramm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge Packungsgröße
18778947 50 μg 50 Mikrogramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 18778947 Lieferant R&D Systems™ Lieferanten-Nr. 8984B7050
 Eingeschränkter Artikel
Verfügbar ab: 03-12-2026
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen

missing translation for 'validMainframeMsg'

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Measured by its ability to induce IFN-gamma secretion by NK-92 human natural killer lymphoma cells. Brandt, C.S. et al. (2009) J. Exp. Med. 206:1495. The ED50 for this effect is 0.5-2.5 μg/mL.
TRUSTED_SUSTAINABILITY

Specifica

Zugriffsnummer XP_005578557
Zur Verwendung mit (Anwendung) Bioactivity
Zusammensetzung Lyophilized from a 0.2 μm filtered solution in PBS.
Gen-ID (Entrez) 374383
Molekulargewicht M.W. (Observed): 70-81 kDa, reducing conditions; M.W. (theoretical): 53 kDa
Menge 50 μg
Quelle Human embryonic kidney cell, HEK293-derived cynomolgus monkey B7-H6 protein Cynomolgus B7-H6 (Asp25-Ser262) (N-terminus) Accession # XP_005578557 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus)
Lagerungsbedingungen Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin-Konzentration <0.10 EU / 1 μg of the protein by the LAL method.
Gen-Alias B7 homolog 6, B7H6, B7-H6, DKFZp686I21167, DKFZp686O24166, NCR3LG1
Gebräuchliche Bezeichnung B7-H6
Gensymbol NCR3LG1
Biologische Aktivität Measured by its ability to induce IFN-gamma secretion by NK-92 human natural killer lymphoma cells. Brandt, C.S. et al. (2009) J. Exp. Med. 206:1495. The ED50 for this effect is typically 0.5-2.5μg/mL.
Konjugat Unconjugated
Spezies Cynomolgus Monkey
Rekombinant Recombinant
Rekonstitution Reconstitute at 100 μg/mL in PBS.
Expressionssystem Human embryonic kidney cell, HEK293-derived Cynomolgus B7-H6 (Asp25-Ser262) Accession # XP_005578557, N-terminus; IEGRMD; Human IgG1 (Pro100-Lys330), C-terminus
Produktlinie R&D Systems
Reinheits- oder Qualitätsgrad >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein B7-H6
Vedi altri risultati Mostra meno risultati
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.