missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Cynomolgus Monkey Axl Fc Chimera Protein
Beschreibung
Measured by its binding ability in a functional ELISA. When Recombinant Cynomolgus Monkey Axl Fc Chimera (Catalog # 11144-AX) is immobilized at 0.25 μg/mL (100 μL/well), Recombinant Human Gas6 (Catalog # 885-GSB ) binds with an ED50 of 0.0700-0.700 ng/mL.
Spezifikation
Spezifikation
| Zugriffsnummer | EHH59605.1 |
| Zur Verwendung mit (Anwendung) | Bioactivity |
| Zusammensetzung | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. |
| Gen-ID (Entrez) | 558 |
| Molekulargewicht | M.W.-observed: 95-105 kDa, under reducing conditions., M.W.-theroretical: 72 kDa |
| Menge | 50 μg |
| Quelle | Chinese Hamster Ovary cell line, CHO-derived cynomolgus monkey Axl protein Cynomolgus Monkey Axl (Glu33-Trp450) (C-terminus) Accession # EHH59605.1 IEGRMD Human IgG1 (Pro100-Lys330) (N-terminus) |
| Lagerungsbedingungen | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. |
| Endotoxin-Konzentration | <0.10 EU / 1 μg of the protein by the LAL method. |
| Gen-Alias | AI323647, ARK, AXL oncogene, AXL receptor tyrosine kinase, AXL transforming sequence/gene, EC 2.7.10, EC 2.7.10.1, JTK11, Tyro7, tyrosine-protein kinase receptor UFO, UFO, UFOoncogene AXL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?