missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Human alcohol dehydrogenase 7 GST (N-Term) Protein
Alle ansehen Bio Techne Produkte
Click to view available options
Menge:
10 μg
25 μg
Packungsgröße:
10 Mikrogramm
25 Mikrogramm
Beschreibung
This protein was produced in an in vitro wheat germ expression system that should preserve correct conformational folding that is necessary for biological function. While it is possible that this protein could display some level of activity, the functionality of this protein has not been explicitly measured or validated.
Spezifikation
Spezifikation
| Zur Verwendung mit (Anwendung) | Western Blot, ELISA, Protein Array, Immunoaffinity Purification |
| Zusammensetzung | 50 mM Tris-HCl, 10 mM reduced Glutathione, pH 8.0 in the elution buffer. |
| Gen-ID (Entrez) | 131 |
| Molekulargewicht | M.W. (Theoretical): 67.9 kDa |
| Menge | 10 μg |
| Lagerungsbedingungen | Store at -80C. Avoid freeze-thaw cycles. |
| Gen-Alias | ADH4, ADH-4, alcohol dehydrogenase 7 (class IV), mu or sigma polypeptide, alcohol dehydrogenase class 4 mu/sigma chain, Alcohol dehydrogenase class IV mu/sigma chain, alcohol dehydrogenase VII, alcohol dehydrogenase-7, class IV sigma-1 alcohol dehydrogenase, class IV sigmasigma alcohol dehydrogenase, EC 1.1.1, EC 1.1.1.1, Gastric alcohol dehydrogenase, Retinol dehydrogenase |
| Gensymbol | ADH7 |
| Reinheits- oder Qualitätsgrad | >80% by SDS-PAGE and Coomassie blue staining |
| Protein | alcohol dehydrogenase 7 |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur