missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human Angiopoietin-like 7 Biotin Protein

Artikelnummer. 18757518
Ansicht ändern
Click to view available options
Menge:
25 μg
Packungsgröße:
25 Mikrogramm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge Packungsgröße
18757518 25 μg 25 Mikrogramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 18757518 Lieferant R&D Systems™ Lieferanten-Nr. BT914
 Eingeschränkter Artikel
Verfügbar ab: 27-11-2026
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen

missing translation for 'validMainframeMsg'

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Human Angiopoietin-like 7 Biotin Protein is derived from CHO. The Recombinant Human Angiopoietin-like 7 Biotin Protein has been validated for the following applications: Bioactivity.
TRUSTED_SUSTAINABILITY

Specifications

Zugriffsnummer O43827
Zusammensetzung Lyophilized from a 0.2μm filtered solution in PBS and NaCl
Gen-ID (Entrez) 10218
Molekulargewicht 39kDa (unlabeled)
Name Angiopoietin-like Protein 7/ANGPTL7
Vorbereitungsmethode Reconstitute at 100μg/mL in PBS.
Qualitätskontrollen SDS-PAGE: >110kDa, reducing conditions
Menge 25 μg
Quelle Chinese Hamster Ovary cell line,CHO-derived human Angiopoietin-like Protein 7/ANGPTL7 protein Gln27-Pro346,with a C-terminal 10-His tag
Lagerungsbedingungen Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Regulatorischer Status RUO
Kündigung No results: Gln27 predicted, sequencing might be blocked
Strukturform Oligomer, Biotinylated on protein carbohydrates
Konjugat Biotin
Rekombinant Recombinant
Produktlinie R&D Systems
Reinheits- oder Qualitätsgrad 95%, by SDS-PAGE with silver staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.