missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human CD19 Fc Chimera Alexa Fluor™ 488 Protein
Click to view available options
Menge:
20 μg
Packungsgröße:
20 Mikrogramm
Beschreibung
Measured by flow cytometry for its ability to bind anti-Human CD19 Monoclonal Antibody conjugated beads. The concentration of Recombinant Human CD19 Fc Chimera Alexa Fluor™ 488 (Catalog # AFG9269) that produces 50% of the binding is 0.10-1.50 μg/mL.
Spezifikation
Spezifikation
| Zugriffsnummer | P15391.6 |
| Zur Verwendung mit (Anwendung) | Bioactivity |
| Zusammensetzung | Supplied as a 0.2 μm filtered solution in PBS with BSA as a carrier protein. |
| Gen-ID (Entrez) | 930 |
| Molekulargewicht | M.W.-observed: 77-92 kDa, reducing conditions, M.W.-theroretical: 57 kDa |
| Menge | 20 μg |
| Quelle | Chinese Hamster Ovary cell line, CHO-derived human CD19 protein Human CD19 (Glu21-Lys291) (N-terminus) Accession # P15391.6 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus) |
| Lagerungsbedingungen | Protect from light. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after opening. 3 months, -20°C to -70°C under sterile conditions after opening. |
| Endotoxin-Konzentration | <1.0 EU / 1 μg of the protein by the LAL method. |
| Gen-Alias | B4, B-lymphocyte antigen CD19, B-lymphocyte surface antigen B4, CD19, CD19 antigen, CD19 molecule, CVID3, Differentiation antigen CD19, Leu-12, MGC12802, T-cell surface antigen Leu-12 |
| Mehr anzeigen |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur