missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human CRACC/SLAMF7 His Protein

Artikelnummer. 18159829 Alle ansehen Bio Techne Produkte
Change view
Click to view available options
Menge:
0,05 mg
0,25 mg
Packungsgröße:
0.05 Milligramm
0.25 Milligramm
Artikelnummer. Menge unitSize
18159829 0,05 mg 0.05 Milligramm
18205020 0,25 mg 0.25 Milligramm
2 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Artikelnummer. 18159829

Marke: Novus Biologicals™ NBP2522580.05MG

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

A recombinant protein with a C-Terminal His-tag and corresponding to the amino acids 23-226 of Human CRACC/SLAMF7 The Recombinant Human CRACC/SLAMF7 Protein is derived from Baculovirus. The Recombinant Human CRACC/SLAMF7 Protein has been validated for the following applications: SDS-Page.
TRUSTED_SUSTAINABILITY

Spezifikation

Konzentration 0.5mg/mL
Zur Verwendung mit (Anwendung) SDS-PAGE
Zusammensetzung Phosphate buffered saline (pH 7.4) containing 10% glycerol
Gen-ID (Entrez) 57823
Molekulargewicht TMW: 23.4kDa
Menge 0,05 mg
Quelle Baculovirus
Lagerungsbedingungen Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles.
Gen-Alias 19A, CD2 subset 1, CD2-like receptor activating cytotoxic cells, CD319, CD319 antigen, CRACCCD2-like receptor-activating cytotoxic cells, CS119A24 protein, Membrane protein FOAP-12, Novel Ly9, novel LY9 (lymphocyte antigen 9) like protein, Protein 19A, SLAM family member 7
Gebräuchliche Bezeichnung CRACC/SLAMF7
Konjugat Unconjugated
Rekombinant Recombinant
Reinheits- oder Qualitätsgrad >95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain
Show More Show Less

For research use only and is not approved for use in humans or in clinical diagnosis

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.