missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Human FGF acidic/FGF1 Protein
Alle ansehen Bio Techne Produkte
Click to view available options
Menge:
10 μg
100 μg
1000 μg
500 μg
Packungsgröße:
10 Mikrogramm
100 Mikrogramm
1000 Mikrogramm
500 Mikrogramm
Beschreibung
FGF acidic/FGF1 protein is fully biologically active when compared to standard. The ED50 as determined by a cell proliferation assay using murine balb/c 3T3 cells is less than 0.5 ng/mL, corresponding to a specific activity of > 2.0 x 10^6 IU/mg in the presence of 10 μg/mL of heparin.
Spezifikation
Spezifikation
| Zur Verwendung mit (Anwendung) | Western Blot, Bioactivity |
| Zusammensetzung | Lyophilized from a 0.2 μm filtered concentrated solution in PBS, pH 7.4, 2 mM EDTA, 0.5 mM DTT. |
| Gen-ID (Entrez) | 2246 |
| Molekulargewicht | MolecularWeight-theroretical: 17.3 kDa |
| Menge | 10 μg |
| Lagerungsbedingungen | Store at -20°C to -70°C as supplied. After reconstitution, store at 2 to 8 C for 1 month and (at -20°C to -70°C for long term storage. Avoid repeated freeze/thaw cycles.) |
| Endotoxin-Konzentration | Less than 0.1 EU/μg of FGF acidic/FGF1 as determined by LAL method. |
| Gen-Alias | AFGF, alpha, ECGF, ECGFB, ECGF-betaAcidic fibroblast growth factor, endothelial cell growth factor, beta, FGFABeta-endothelial cell growth factor, FGF-alpha, fibroblast growth factor 1 (acidic), GLIO703, HBGF1, HBGF-1, heparin-binding growth factor 1 |
| Gensymbol | FGF1 |
| Rekonstitution | Recommended to centrifuge prior to opening. Reconstitute in sterile distilled water or aqueous buffer containing 0.1% BSA to a concentration of 0.1-1.0 mg/mL Apportion stock solutions into working aliquots and store (at <-20°C.) |
| Mehr anzeigen |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur