missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human IgG1 Fc Chimera Alexa Fluor™ 488 Protein
Click to view available options
Menge:
20 μg
Packungsgröße:
20 Mikrogramm
Beschreibung
Measured by flow cytometry for its ability to bind anti-human IgG1 Fc Antibody conjugated beads. The concentration of Recombinant Human IgG1Fc Chimera Alexa Fluor™ 647 (Catalog # AFR110) that produces 50% of the binding response is 10.0-100 ng/mL.
Spezifikation
Spezifikation
| Zugriffsnummer | P01857 |
| Zusammensetzung | Supplied as a 0.2 μm filtered solution in PBS with BSA as a carrier protein. |
| Molekulargewicht | M.W.-observed: 30-40 kDa, under reducing conditions., M.W.-theroretical: 26.6 kDa |
| Menge | 20 μg |
| Quelle | Mouse myeloma cell line, NS0-derived human IgG1 protein IEGRMD Human IgG1 Pro100-Lys330 (Leu117Ala, Leu118Glu, Gly120Ala, Thr276Ala) (C-terminus) Accession # P01857 (N-terminus) |
| Lagerungsbedingungen | Protect from light. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after opening. 3 months, -20°C to -70°C under sterile conditions after opening. |
| Endotoxin-Konzentration | <1.0 EU / 1 μg of the protein by the LAL method. |
| Gen-Alias | IgG1, Igh-4, RGD1359539, VH7183 |
| Konjugat | Alexa Fluor 488 |
| Reinheits- oder Qualitätsgrad | >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining. |
| Mehr anzeigen |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur