Learn More
R&D Systems™ Recombinant Human IL-1 beta/IL-1F2 Protein
Beschreibung
Measured in a cell proliferation assay using D10.G4.1 mouse helper T cells. Symons, J.A. et al. (1987) in Lymphokines and Interferons, a Practical Approach. Clemens, M.J. et al. (eds): IRL Press. 272. The ED50 for this effect is <12 pg/mL. The specific activity of recombinant human IL-1 beta is approximately 1.4 x 105 IU/μg, which is calibrated against recombinant human IL-1 beta WHO International Standard (NIBSC code: 86/680). Specific activity is for reference purposes only and is not routinely tested.
Spezifikation
Spezifikation
| Zusammensetzung | Lyophilisiert aus einer 0,2 μm gefilterten Lösung in PBS mit Trehalose. Packungsgröße mit 1 mg (01M) wird als 0,2 μm gefilterte Lösung in PBS geliefert. Einzelheiten finden Sie im Analysezertifikat. |
| Gen-ID (Entrez) | 3553 |
| Molekulargewicht | 17 kDa |
| Name | IL-1 beta/IL-1F2 |
| Menge | 5 μg |
| Quelle | E. coli-derived human IL-1 beta/IL-1F2 protein Ala117-Ser269 |
| Lagerungsbedingungen | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution. The 1 mg pack size (01M) can be stored for 1 month at 2 to 8 °C under sterile conditions after opening. |
| Gen-Alias | catabolin, IL-1, IL1 beta, IL-1 beta, IL1B, IL-1B, IL1-BETA, IL-1F2, IL1F2IL-1 beta, interleukin 1, beta, interleukin-1 beta, preinterleukin 1 beta, pro-interleukin-1-beta |
| Konjugat | Unconjugated |
| Rekombinant | Recombinant |
| Mehr anzeigen |