missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human N-glycosylase / DNA lyase Protein

Artikelnummer. 16083370
Click to view available options
Menge:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
1 Milligramm
10 Mikrogramm
100 Mikrogramm
200 Mikrogramm
50 Mikrogramm
500 Mikrogramm
This item is not returnable. View return policy

Product Code. 16083370

Brand: enQuireBio™ QP6448ec100ug

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

This item is not returnable. View return policy

A DNA sequence encoding the Homo sapiens (Human) N-glycosylase/DNA lyase, was expressed in the hosts and tags indicated.

Specifications

Zugriffsnummer NP_002533.1
Gen-ID (Entrez) 4968
Name N-glycosylase / DNA lyase Protein
Menge 100 μg
Kennzeichnung Research Use Only
Endotoxin-Konzentration Not determined.
Gensymbol OGG1
Proteinlänge Full Length
Produkttyp Recombinant Protein
Kreuzreaktivität Human
Spezies E. coli
Proteinmarkierung His
Sequenz MPARALLPRR MGHRTLASTP ALWASIPCPR SELRLDLVLP SGQSFRWREQ SPAHWSGVLA DQVWTLTQTE EQLHCTVYRG DKSQASRPTP DELEAVRKYF QLDVTLAQLY HHWGSVDSHF QEVAQKFQGV RLLRQDPIEC LFSFICSSNN NIARITGMVE RLCQAFGPRL IQLDDVTYHG FPSLQALAGP EVEAHLRKLG LGYRARYVSA SARAILEEQG GLAWLQQLRE SSYEEAHKAL CILPGVGTKV ADCICLMALD KPQAVPVDVH MWHIAQRDYS WHPTTSQAKG PSPQTNKELG NFFRSLWGPY AGWAQAVLFS ADLRQSRHAQ EPPAKRRKGS KGPEG
Puffer Tris-based buffer, 50% glycerol
Reinheits- oder Qualitätsgrad Greater than 90% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.