Learn More
Novus Biologicals™ Recombinant Human Neuregulin-1 beta 1/NRG1 beta 1 Protein
Artikelnummer. 18476711 Alle ansehen Bio Techne Produkte
Change view
Click to view available options
Menge:
1 mg
10 μg
100 μg
500 μg
Packungsgröße:
1 Milligramm
10 Mikrogramm
100 Mikrogramm
500 Mikrogramm
Artikelnummer. Menge unitSize
18476711 10 μg 10 Mikrogramm
18427261 100 μg 100 Mikrogramm
18410152 1 mg 1 Milligramm
18459491 500 μg 500 Mikrogramm
4 options
Artikelnummer. 18476711
Marke: Novus Biologicals™ NBP23509710ug
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Recombinant Protein
Neuregulin-1 beta 1/NRG1 beta 1 protein is fully biologically active when compared to standard. The ED50 as determined by a cell proliferation assay using serum free human MCF-7 cells is less than 0.5 ng/mL, corresponding to a specific activity of >2.0 x 106IU/mg.
TRUSTED_SUSTAINABILITY
Spezifikation
Zugriffsnummer Q02297, Q02297, Q02297
Konzentration LYOPH
Zur Verwendung mit (Anwendung) PAGE, Bioactivity
Zusammensetzung Lyophilized from a 0.2 um filtered solution in 1 x PBS, pH 7.4.
Gen-ID (Entrez) 3084
Molekulargewicht 7.5 kDa
Menge 10 μg
Lagerungsbedingungen Store at -20°C to -70°C as supplied. After reconstitution, store at 2°C to 8°C for 1 month and at -20°C to -70°C for long term storage. Avoid repeated freeze-thaw cycles.
Endotoxin-Konzentration Less than 0.1 EU/ug of Neuregulin-1 beta 1/NRG1 beta 1 as determined by LAL method.
Gen-Alias ARIA, GGF, GGF2, HGL, HRG, HRG1, HRGA, MST131, MSTP131, NDF, neuregulin 1, NRG1-IT2, SMDF
Gensymbol NRG1
Rekonstitution Recommended to centrifuge prior to opening. Reconstitute in sterile distilled water or aqueous buffer containing 0.1% BSA to a concentration of 0.1-1.0mg/mL. Apportion stock solutions into working aliquots and store at <-20°C.
Reinheits- oder Qualitätsgrad >97% pure by SDS-PAGE and HPLC
Protein Neuregulin-1 beta 1/NRG1 beta 1