missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Nuclear pore membrane glycoprotein 210 Protein

Artikelnummer. 16083330
Click to view available options
Menge:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Packungsgröße:
1 Milligramm
10 Mikrogramm
100 Mikrogramm
200 Mikrogramm
50 Mikrogramm
500 Mikrogramm
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Artikelnummer. 16083330

Marke: enQuireBio™ QP6440ec1mg

om dit product te kopen Registreer vandaag om een webaccount aan te maken

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

A DNA sequence encoding the Homo sapiens (Human) Nuclear pore membrane glycoprotein 210, was expressed in the hosts and tags indicated.

Specifications

Zugriffsnummer NP_079199.2
Gen-ID (Entrez) 23225
Name Nuclear pore membrane glycoprotein 210 Protein
Menge 1 mg
Kennzeichnung Research Use Only
Endotoxin-Konzentration Not determined.
Gensymbol NUP210
Proteinlänge Partial (see sequence information for more details).
Produkttyp Recombinant Protein
Kreuzreaktivität Human
Spezies E. coli
Proteinmarkierung His
Sequenz AVGSVTVYYE VAGHLRTYKE VVVSVPQRIM ARHLHPIQTS FQEATASKVI VAVGDRSSNL RGECTPTQRE VIQALHPETL ISCQSQFKPA VFDFPSQDVF TVEPQFDTAL GQYFCSITMH RLTDKQRKHL SMKKTALVVS ASLSSSHFST EQVGAEVPFS PGLFADQAEI LLSNHYTSSE IRVFGAPEVL ENLEVKSGSP AVLAFAKEKS FGWPSFITYT VGVLDPAAGS QGPLSTTLTF SSPVTNQAIA IPVTVAFVVD RRGPGPYGAS LFQHFLDSYQ
Puffer Tris-based buffer, 50% glycerol
Reinheits- oder Qualitätsgrad Greater than 90% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.