missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Protachykinin-1 Protein

Artikelnummer. 16075610
Change view
Click to view available options
Menge:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Conditionnement:
1 Milligramm
10 Mikrogramm
100 Mikrogramm
200 Mikrogramm
50 Mikrogramm
500 Mikrogramm
Product Code. Menge unitSize
16075610 1 mg 1 Milligramm
16065610 10 μg 10 Mikrogramm
16005620 50 μg 50 Mikrogramm
16055610 100 μg 100 Mikrogramm
16085610 200 μg 200 Mikrogramm
16095610 500 μg 500 Mikrogramm
6 options
This item is not returnable. View return policy

Product Code. 16075610

Brand: enQuireBio™ QP6758ec1mg

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

This item is not returnable. View return policy

A DNA sequence encoding the Homo sapiens (Human) Protachykinin-1, was expressed in the hosts and tags indicated.

Specifications

Zugriffsnummer NP_003173.1
Gen-ID (Entrez) 6863
Name Protachykinin-1 Protein
Menge 1 mg
Kennzeichnung Research Use Only
Endotoxin-Konzentration Not determined.
Gensymbol TAC1
Proteinlänge Full Length
Produkttyp Recombinant Protein
Kreuzreaktivität Human
Spezies E. coli
Proteinmarkierung GST
Sequenz MKILVALAVF FLVSTQLFAE EIGANDDLNY WSDWYDSDQI KEELPEPFEH LLQRIARRPK PQQFFGLMGK RDADSSIEKQ VALLKALYGH GQISHKRHKT DSFVGLMGKR ALNSVAYERS AMQNYERRR
Puffer Tris-based buffer, 50% glycerol
Reinheits- oder Qualitätsgrad Greater than 80% as determined by SDS-PAGE.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.